Learn More
Mopec Sink Cabinet, ADA Compliant Approach
SDP
Catalog No. 21200373 Shop All Mopec Products
Change view
Click to view available options
Type:
Compartment Sink
Catalog No. Type
21-200-373 Compartment Sink
1 options
Catalog No. 21-200-373 Supplier Mopec Supplier No. LC981ACC36
Add to Cart
Add to Cart
Mopec’s Casework Line offers durable, modular stainless-steel cabinetry for surgical pathology, autopsy, and lab settings. Built for longevity, easy cleaning, and seamless integration, it supports efficient, organized, and ergonomic workflows in clinical environments.
Construction
  • ADA Height Cabinets are 32.625 in. tall with leveling bolt zeroed out and 22 in. deep
  • This cabinet is meant for a person in a wheelchair to be able to roll up to/under a sink
  • Sink supports are sold separately
Materials
  • 304 stainless steel construction with a polished No. 4 finish
Dimensions
  • LC981ACC: 36 in. L x 22 in. W x 32.6 in. H
Specifications
Color Light Gray
Material Stainless Steel
Type Compartment Sink