Learn More
Mopec Sink Cabinet, Standing Height, 1 Door
SDP
Catalog No. 21720006 Shop All Mopec Products
Change view
Click to view available options
Type:
Compartment Sink
Catalog No. Type
21-720-006 Compartment Sink
1 options
Catalog No. 21-720-006 Supplier Mopec Supplier No. LC19424
Add to Cart
Add to Cart
Mopec’s Casework Line offers durable, modular stainless-steel cabinetry for surgical pathology, autopsy, and lab settings. Built for longevity, easy cleaning, and seamless integration, it supports efficient, organized, and ergonomic workflows in clinical environments.
Construction
  • (1) Standing Height Cabinet with leveling bolt zeroed out
  • (1) Left Hinge Door
  • Sink Supports are sold separately
Materials
  • 304 stainless steel construction with a polished No. 4 finish
Dimensions
  • LC194-24: 24 in. L x 22 in. W x 35.1 in. H
Compatibility
  • Cabinets are compatible with LC535 sink support kits
Specifications
Color Light Gray
Material Stainless Steel
Type Compartment Sink