Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dynamic Diagnostics Sink Splash Guard with Easy Cut-Away

Catalog No. 22899673
Change view
Click to view available options
Length (English):
18.75 in.
23.625 in.
Dimensions (L x W x H):
18.75 x 4.5 x 18 in.
23.625 x 4.5 x 18 in.
Size:
Small
Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Dimensions (L x W x H) Size
22-899-673 23.625 in. 23.625 x 4.5 x 18 in. Medium
22-899-672 18.75 in. 18.75 x 4.5 x 18 in. Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-899-673 Supplier Dynamic Diagnostics Supplier No. 4000149
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Bio hazard safety shield

  • Easy cut-away

Specifications

Width (English) 4.5 in.
Color Clear
Depth (English) 18 in.
Length (English) 23.625 in.
Dimensions (L x W x H) 23.625 x 4.5 x 18 in.
Size Medium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.