Learn More
Foxx Life Sciences EZBio Dip Tube Sinker, PVDF, 1/2" Barb, Individually Double Bagged, Non-Sterile , 5/CS
Catalog No. 502060586
Encompass_Preferred
Foxx Life Sciences offers a premium line of high-quality PVDF Anchor dip tube sinkers. The Anchor is developed with you in mind to help eliminate the irritation of moving dip tubes. With this unique design, the Anchor helps to ensure optimal depth and flow rate, while removing the risk of suction against all containers. SpecificationsAnchor Material: PVDFBarb Size: 1/2"Part Diameter: 1.00"Gamma Irradiated: NoQuantity: Pack of 5Packaging: Individually Double Bagged
Catalog No. 50-206-0586 Supplier Foxx Life Sciences Supplier No. 25S1236FLS
Foxx Life Sciences offers a premium line of high-quality PVDF Anchor dip tube sinkers. The Anchor is developed with you in mind to help eliminate the irritation of moving dip tubes. With this unique design, the Anchor helps to ensure optimal depth and flow rate, while removing the risk of suction against all containers. SpecificationsAnchor Material: PVDFBarb Size: 1/2"Part Diameter: 1.00"Gamma Irradiated: NoQuantity: Pack of 5Packaging: Individually Double Bagged