Learn More
RWD LIFE SCIENCE INC SINLGE CELL SUSP DISSOCIATOR
SDP
Supplier:  RWD LIFE SCIENCE INC DSC410
Encompass_Preferred
RWD Single-cell suspension dissociator uses tissue processing tubes, tissue dissociation kits, and built-in optimization programs for preparing high-activity single-cell suspension and tissue homogenate. The preparation of single-cell suspension with high cell viability can be completed within 15-30 minutes, which makes the experiments more reproducible. 4 independent operating channels and heating jackets can automate the processes and improve the efficiency of tissue processing.Heating jackets are also available. Dissociator with heaters will save you from time-consuming water bath and improve efficiency. Moreover, all the parameters can be adjusted in user-customized programs to process more kinds of tissue types.
Catalog No. NC3404363
Add to cart
Request a Quote