Learn More
Sino Biological Sinofection Transfection Reagent 1 mL
Supplier:  Sino Biological STF021
Encompass_Preferred
Sinofection is a superior cationic polymer-based transfection reagent. It has been successfully applied to transfection at various scales over a broad range of cell lines. Compared with the leading transfection reagents on the market, Sinofection yields higher protein expression level and exhibits lower cytotoxicity. It is an ideal choice for recombinant protein production for both transient and stable transfection. Its extensive compatibility with serum, cell type, scale, and storage temperature offers you ease and assurance for research and production.
Catalog No. 50-006-96285