Learn More
Medchemexpress LLC Sinomenine | 115-53-7 | C19H23NO4 | 100 MG
SDP
Catalog No. 5000375755
Encompass_Preferred
Sinomenine is an alkaloid extracted from *Sinomenium acutum*. It functions as a blocker of NF-κB activation and also acts as an activator of the μ-opioid receptor.
  • Source: extracted from the plant *Sinomenium acutum*.
  • NF-κB blocker: inhibits NF-κB activation.
  • Μ-Opioid receptor activator: activates the μ-opioid receptor.
  • Antiproliferative activity: demonstrated antiproliferative activity against various human cancer cell lines, including K562.
  • Anti-inflammatory and analgesic effects: shows anti-inflammatory activity, for instance, by inhibiting LPS-induced nitric oxide production in RAW264.7 cells. In rodents, it produces antinociception in hot plate and tail flick tests and reduces mechanical hypersensitivity in nerve-injured mice.
  • Research use: primarily for research purposes.
Catalog No. 50-003-75755 Supplier Medchemexpress LLC Supplier No. HY15122100MG
Sinomenine is an alkaloid extracted from *Sinomenium acutum*. It functions as a blocker of NF-κB activation and also acts as an activator of the μ-opioid receptor.
  • Source: extracted from the plant *Sinomenium acutum*.
  • NF-κB blocker: inhibits NF-κB activation.
  • Μ-Opioid receptor activator: activates the μ-opioid receptor.
  • Antiproliferative activity: demonstrated antiproliferative activity against various human cancer cell lines, including K562.
  • Anti-inflammatory and analgesic effects: shows anti-inflammatory activity, for instance, by inhibiting LPS-induced nitric oxide production in RAW264.7 cells. In rodents, it produces antinociception in hot plate and tail flick tests and reduces mechanical hypersensitivity in nerve-injured mice.
  • Research use: primarily for research purposes.