Learn More
Apexbio Technology LLC Sinomenine Hydrochloride 6080-33-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000090099
Encompass_Preferred
Sinomenine Hydrochloride (CAS 6080-33-7) is an alkaloid derivative isolated from the stems of Sinomenium acutum It acts primarily by modulating the immune response through inhibition of pro-inflammatory cytokine production and suppression of NF- B signaling pathways Sinomenine Hydrochloride has been used in preclinical studies to investigate its anti-inflammatory and immunosuppressive activities particularly in models of rheumatoid arthritis and other autoimmune conditions This compound is suited for use in biomedical research exploring inflammation immune modulation and related therapeutic mechanisms Store in a sealed container under cool dry conditions
Catalog No. 50-000-90099 Supplier Apexbio Technology LLC Supplier No. N21895.1
Sinomenine Hydrochloride (CAS 6080-33-7) is an alkaloid derivative isolated from the stems of Sinomenium acutum It acts primarily by modulating the immune response through inhibition of pro-inflammatory cytokine production and suppression of NF- B signaling pathways Sinomenine Hydrochloride has been used in preclinical studies to investigate its anti-inflammatory and immunosuppressive activities particularly in models of rheumatoid arthritis and other autoimmune conditions This compound is suited for use in biomedical research exploring inflammation immune modulation and related therapeutic mechanisms Store in a sealed container under cool dry conditions