Learn More
STEMCELL Technologies Sinomenine (Hydrochloride), Size: 50 mg
SDP
Supplier:  STEMCELL Technologies 72882
Encompass
Sinomenine is a natural plant alkaloid commonly used to alleviate inflammation associated with rheumatoid arthritis (Wang and Li). It impairs signaling through Nuclear factor-kappaB (NF-κB; Sun et al.; Wang and Li) and enhances the bioavailability of some compounds, at least in part through an inhibition of drug export by transporters like P-glycoprotein (Kesarwani et al.; Liu et al.).MAINTENANCE AND SELF-RENEWAL· Promotes self-renewal in cultured human and mouse embryonic stem cells (Desbordes et al.).
Catalog No. 50-197-5995
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov