Learn More
Cayman Chemical Sinomenine (hydrochloride), Purity: ≥98%, CAS: 6080-33-7, 100 mg
SDP
Catalog No. 502971691
Encompass_Preferred
A natural plant alkaloid commonly used to alleviate inflammation associated with rheumatoid arthritis; activates histamine release, reduces joint stiffness and pain, and alters cytokine generation; impairs signaling through NF-κB, resulting in immunosuppression as well as reduced inflammation and pain
Catalog No. 50-297-1691 Supplier Cayman Chemical Supplier No. 16143100 MG
A natural plant alkaloid commonly used to alleviate inflammation associated with rheumatoid arthritis; activates histamine release, reduces joint stiffness and pain, and alters cytokine generation; impairs signaling through NF-κB, resulting in immunosuppression as well as reduced inflammation and pain