Learn More
Hach Company Replacement SIP 10 Sipper Unit for use with DR 3900 Spectrophotometer
Catalog No. 501997830 Shop All Hach Company Products
Encompass
Replacement SIP 10 Module (Sipper module only). This part does not include the required flow cell, tubing or USB cable. For use with the DR 3900 Spectrophotometer. For Complete Sipper set up, please order LQV157.99.10002.
Catalog No. 50-199-7830 Supplier Hach Company Supplier No. LQV157.99.00002
Replacement SIP 10 Module (Sipper module only). This part does not include the required flow cell, tubing or USB cable. For use with the DR 3900 Spectrophotometer. For Complete Sipper set up, please order LQV157.99.10002.