Learn More
Hach Company SIP10 Sipper Kit with Tubing and Dual Path Length Cell
Catalog No. 501997831 Shop All Hach Company Products
Encompass
The SIP 10 sipper system works with the DR 3900 spectrophotometer for pour thru applications, including Hach methods. It is only compatible with the DR 3900. The sipper system is powered via USB connection to the DR 3900 and is controlled through the DR 3900 menu.
Catalog No. 50-199-7831 Supplier Hach Company Supplier No. LQV157.99.10002
The SIP 10 sipper system works with the DR 3900 spectrophotometer for pour thru applications, including Hach methods. It is only compatible with the DR 3900. The sipper system is powered via USB connection to the DR 3900 and is controlled through the DR 3900 menu.