Learn More
Hach Company SIP 10 Sipper Set for pour-thru methods, 1 inch vial
Catalog No. 501997832 Shop All Hach Company Products
Encompass
The SIP 10 sipper system works with the DR 6000 spectrophotometer for pour thru applications, including Hach methods. It is only compatible with the DR 6000. The sipper system is powered via USB connection to the DR6000 and is controlled through the DR 6000 menu.
Catalog No. 50-199-7832 Supplier Hach Company Supplier No. LQV157.99.20002
The SIP 10 sipper system works with the DR 6000 spectrophotometer for pour thru applications, including Hach methods. It is only compatible with the DR 6000. The sipper system is powered via USB connection to the DR6000 and is controlled through the DR 6000 menu.