Learn More
Aviva Systems Biology Corp SIP1 Antibody - middle region (ARP40272_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40272_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SIP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SIP1. Alternate names: GEMIN2, SIP1-delta, SIP1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57372