Learn More
Hach Company SIP10 Sipper Unit TU5200
Catalog No. 5000207551 Shop All Hach Company Products
Encompass
SIP10 Sipper Unit for TU5200 Benchtop Laser TurbidimeterPeristaltic pump and lid for pour thru measurement
Catalog No. 50-002-07551 Supplier Hach Company Supplier No. LQV157.99.50002
SIP10 Sipper Unit for TU5200 Benchtop Laser TurbidimeterPeristaltic pump and lid for pour thru measurement