Learn More
AdipoGen Sipholenol A (100 µg)
SDP
Catalog No. 502053689
Encompass_Preferred
Chemical. CAS: 78518-73-7. Formula: C30H52O4. MW: 476.7. Isolated from marine sponge Siphonochalina siphonella. Efficient inhibitor of P-glycoprotein (Pgp, MDR-1) function through direct interactions. By inhibiting MDR-1, active compounds remain in the cytoplasm longer and allowing them to be more effective within the cell. Reversing agent for the treatment of multidrug resistance (MDR) in Pgp-overexpressing tumors. Potentiates the cytotoxicity of established P-gp substrates such as colchicine, vinblastine and paclitaxel. Antiproliferative and cytotoxic compound. Potent antifouling compound.
Catalog No. 50-205-3689 Supplier AdipoGen Supplier No. AGCN20506C100
Add to Cart
Add to Cart
Chemical. CAS: 78518-73-7. Formula: C30H52O4. MW: 476.7. Isolated from marine sponge Siphonochalina siphonella. Efficient inhibitor of P-glycoprotein (Pgp, MDR-1) function through direct interactions. By inhibiting MDR-1, active compounds remain in the cytoplasm longer and allowing them to be more effective within the cell. Reversing agent for the treatment of multidrug resistance (MDR) in Pgp-overexpressing tumors. Potentiates the cytotoxicity of established P-gp substrates such as colchicine, vinblastine and paclitaxel. Antiproliferative and cytotoxic compound. Potent antifouling compound.