Learn More
AdipoGen Sipholenone A (100 µg)
SDP
Supplier:  AdipoGen AGCN20507C100
Encompass_Preferred
Chemical. CAS: 78518-74-8. Formula: C30H50O4. MW: 474.7. Isolated from marine sponge Siphonochalina siphonella. Efficient inhibitor of P-glycoprotein (Pgp, MDR-1) function through direct interactions. By inhibiting MDR-1, active compounds remain in the cytoplasm longer and allowing them to be more effective within the cell. Reversing agent for the treatment of multidrug resistance (MDR) in Pgp-overexpressing tumors. Potentiates the cytotoxicity of established P-gp substrates such as colchicine, vinblastine and paclitaxel. Antiproliferative, antiangiogenic and cytotoxic compound. Moderate antifouling compound.
Catalog No. 50-205-3690