Learn More
Chemglass Life Sciences Siphon Assembly ONLY, for 250mL Media Bottle
SDP
Catalog No. 50143178
Encompass
Siphon Assembly ONLY, for 250mL Media Bottle. Replacement part for 50143213 (250mL media bottle). Siphon assembly consists of stainless steel siphon pipe and silicone tubing cut to length. All stainless parts are manufactured from 316L stainless steel electropolished to a 20Ra or better finish. The silicone tubing is peroxide cured and meets 21 CFR 177.2600 material specifications.Assembled, the media bottle with siphon assembly allows the user to create precisely the desired dip tube angle to retrieve virtually all of the contents of fluid from the bottle.
Catalog No. 50-143-178 Supplier Chemglass Life Sciences Supplier No. CLS1421250
Siphon Assembly ONLY, for 250mL Media Bottle. Replacement part for 50143213 (250mL media bottle). Siphon assembly consists of stainless steel siphon pipe and silicone tubing cut to length. All stainless parts are manufactured from 316L stainless steel electropolished to a 20Ra or better finish. The silicone tubing is peroxide cured and meets 21 CFR 177.2600 material specifications.Assembled, the media bottle with siphon assembly allows the user to create precisely the desired dip tube angle to retrieve virtually all of the contents of fluid from the bottle.