Learn More
Uline Siphon Drum Pump
Catalog No. NC0518844
Encompass_Preferred
Portable hand pump for dispensing mild corrosives, detergents, light oils and waxes, polyethylene, suction tube reaches 34"""" below drum lid, trim to fit any drum, includes adapter to fit 2"""" drum openings
Catalog No. NC0518844 Supplier Uline Supplier No. S17128
Portable hand pump for dispensing mild corrosives, detergents, light oils and waxes, polyethylene, suction tube reaches 34"""" below drum lid, trim to fit any drum, includes adapter to fit 2"""" drum openings