Learn More
Uline SIPHON PAIL PUMP MOQ10
Catalog No. NC2391752
Encompass_Preferred
Minimum order quantity 10. Order in multiples of 10. Priced by "Each"Dispense mild corrosives, detergents, light oils and waxes.Empty a 5-gallon pail in 3 minutes. 21" discharge tube.Polyethylene construction.Suction tube reaches 16" below pail lid.Trim to fit any container.FDA compliant.
Catalog No. NC2391752 Supplier Uline Supplier No. S25309
Minimum order quantity 10. Order in multiples of 10. Priced by "Each"Dispense mild corrosives, detergents, light oils and waxes.Empty a 5-gallon pail in 3 minutes. 21" discharge tube.Polyethylene construction.Suction tube reaches 16" below pail lid.Trim to fit any container.FDA compliant.