Learn More
Medchemexpress LLC Siplizumab, (Synonyms: MEDI-507) 1mg
SDP
Supplier:  Medchemexpress LLC HYP999041MG
Encompass_Preferred
Siplizumab (MEDI-507) is a humanized IgG1 monoclonal antibody against CD2. Siplizumab depletes T cells, decreases T cell activation, inhibites T cell proliferation and enriches naïve and bona fide regulatory T cells.
Catalog No. NC2862463