Learn More
TARGETMOL CHEMICALS INC SIPONIMOD 25MG
SDP
Catalog No. 502579388
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BAF312 (Siponimod (BAF-312)) a next-generation S1P receptor modulator is specific for S1P1 and S1P5 receptors with EC50 of 0.39 nM and 0.98 nM respectively. The specificity of BAF312 for S1P1 and S1P5 receptors exhibits >1000-fold over S1P2 S1P3 and S1P4 receptors. purity: 99%
Catalog No. 50-257-9388 Supplier TARGETMOL CHEMICALS INC Supplier No. T640325MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BAF312 (Siponimod (BAF-312)) a next-generation S1P receptor modulator is specific for S1P1 and S1P5 receptors with EC50 of 0.39 nM and 0.98 nM respectively. The specificity of BAF312 for S1P1 and S1P5 receptors exhibits >1000-fold over S1P2 S1P3 and S1P4 receptors. purity: 99%