Learn More
Medchemexpress LLC HY-12355 5mg , Siponimod CAS:1230487-00-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY123555MG
Encompass_Preferred
HY-12355 5mg Siponimod CAS: 1230487-00-9 Siponimod (BAF-312) is an orally active and selective sphingosine-1-phosphate (S1P) receptor modulator. Siponimod is selective for S1P1 and S1P5 over S1P2, S1P3, and S1P4, with EC50s of 0.4, 0.98, >10000, >1000, and 750 nM, respectively. Siponimod can be used for multiple sclerosis (MS) research-[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0622