Learn More
Cytoskeleton, Inc. SiR-Actin Kit, 50 nmol SiR-Actin and 1 umol verapamil
Catalog No. NC1262651
Encompass
SiR-actin is based on the F-actin binding natural product jasplakinolide. SiR-actin is fluorogenic, cell permeable and highly specific for F-actin. Sir-actin stains endogenous F-actin without the need for genetic manipulation or overexpression. Its emission in the far red minimizes phototoxicity and sample autofluorescence. SiR-actin is compatible with GFP and/or m-cherry fluorescent proteins. It can be imaged with standard Cy5 filtersets. SiR-actin can be used for widefield, confocal, SIM or STED imaging in living cells and tissue. Probe quantity allows 50 – 200 staining experiments.
Catalog No. NC1262651 Supplier Cytoskeleton, Inc. Supplier No. CYSC001
SiR-actin is based on the F-actin binding natural product jasplakinolide. SiR-actin is fluorogenic, cell permeable and highly specific for F-actin. Sir-actin stains endogenous F-actin without the need for genetic manipulation or overexpression. Its emission in the far red minimizes phototoxicity and sample autofluorescence. SiR-actin is compatible with GFP and/or m-cherry fluorescent proteins. It can be imaged with standard Cy5 filtersets. SiR-actin can be used for widefield, confocal, SIM or STED imaging in living cells and tissue. Probe quantity allows 50 – 200 staining experiments.