Learn More
Cytoskeleton, Inc. SIR-LYSOSOME KIT
Catalog No. NC1370956
Encompass
SiR-lysosome Kit (CY-SC012) live cell fluorogenic lysosome labeling probe
Catalog No. NC1370956 Supplier Cytoskeleton, Inc. Supplier No. CYSC012
SiR-lysosome Kit (CY-SC012) live cell fluorogenic lysosome labeling probe