Learn More
Cytoskeleton, Inc. SiR-NHS Ester, 1mg, SiR-NHS ester is the N-hyroxysuccinimidyl ester of SiR
Catalog No. NC2948718
Encompass
SiR-NHS ester is the N-hyroxysuccinimidyl ester of SiR. This amine reactive probe enables the preparation of custom SiR probes. SiR-NHS ester can be coupled to any amine bearing ligand: antibody, drug, protein or nucleic acid. SiR is compatible with GFP and/or m-cherry fluorescent proteins. It can be imaged with standard Cy5 filtersets.
Catalog No. NC2948718 Supplier Cytoskeleton, Inc. Supplier No. CYSC003
SiR-NHS ester is the N-hyroxysuccinimidyl ester of SiR. This amine reactive probe enables the preparation of custom SiR probes. SiR-NHS ester can be coupled to any amine bearing ligand: antibody, drug, protein or nucleic acid. SiR is compatible with GFP and/or m-cherry fluorescent proteins. It can be imaged with standard Cy5 filtersets.