Learn More
Cytoskeleton, Inc. SiR-Tubulin Kit, 50 nmol SiR-Tubulin and 1 umol verapamil
Catalog No. NC0958386
Encompass
SiR-tubulin is based on the microtubule binding drug Docetaxel. SiR-tubulin is fluorogenic, cell permeable and highly specific for microtubules. Sir-tubulin stains endogenous microtubules without the need for genetic manipulation or overexpression. Its emission in the far red minimizes phototoxicity and sample autofluorescence. SiR-tubulin is compatible with GFP and/or m-cherry fluorescent proteins. It can be imaged with standard Cy5 filtersets. SiR-tubulin can be used for widefield, confocal, SIM or STED imaging in living cells and tissue. Probe quantity allows 50 – 200 staining experiments.
Catalog No. NC0958386 Supplier Cytoskeleton, Inc. Supplier No. CYSC002
SiR-tubulin is based on the microtubule binding drug Docetaxel. SiR-tubulin is fluorogenic, cell permeable and highly specific for microtubules. Sir-tubulin stains endogenous microtubules without the need for genetic manipulation or overexpression. Its emission in the far red minimizes phototoxicity and sample autofluorescence. SiR-tubulin is compatible with GFP and/or m-cherry fluorescent proteins. It can be imaged with standard Cy5 filtersets. SiR-tubulin can be used for widefield, confocal, SIM or STED imaging in living cells and tissue. Probe quantity allows 50 – 200 staining experiments.