Learn More
Medchemexpress LLC Siramesine (hydrochloride) | 224177-60-0 | 99.3% | C30H32ClFN2O | 100 MG
SDP
Catalog No. 5000356907
Encompass_Preferred
Siramesine hydrochloride (Lu 28-179) is a potent sigma-2 receptor agonist with subnanomolar affinity (IC50=0.12 nM) and 140-fold selectivity for sigma-2 receptors over sigma-1 receptors (IC50=17 nM). It induces cell death by destabilizing mitochondria and exhibits anti-cancer activity.
  • Potent sigma-2 receptor agonist
  • Displays subnanomolar affinity for sigma-2 receptors
  • Induces cell death by destabilizing mitochondria
  • Exhibits anti-cancer activity
  • Activates caspases in HaCaT and U-87MG cells
Catalog No. 50-003-56907 Supplier Medchemexpress LLC Supplier No. HY14221A100MG
Siramesine hydrochloride (Lu 28-179) is a potent sigma-2 receptor agonist with subnanomolar affinity (IC50=0.12 nM) and 140-fold selectivity for sigma-2 receptors over sigma-1 receptors (IC50=17 nM). It induces cell death by destabilizing mitochondria and exhibits anti-cancer activity.
  • Potent sigma-2 receptor agonist
  • Displays subnanomolar affinity for sigma-2 receptors
  • Induces cell death by destabilizing mitochondria
  • Exhibits anti-cancer activity
  • Activates caspases in HaCaT and U-87MG cells