Learn More
Medchemexpress LLC Siramesine hydrochloride | 224177-60-0 | 99.7% | C30H32ClFN2O | 2 MG
SDP
Catalog No. 5000353299
Encompass_Preferred
Siramesine hydrochloride, also known as Lu 28-179 hydrochloride, is a potent sigma-2 receptor agonist. It demonstrates high selectivity for sigma-2 receptors over sigma-1 receptors. This compound triggers cell death by destabilizing mitochondria and exhibits anti-cancer activity.
  • Potent sigma-2 receptor agonist
  • High selectivity for sigma-2 receptors
  • Triggers cell death via mitochondrial destabilization
  • Shows anti-cancer activity
Catalog No. 50-003-53299 Supplier Medchemexpress LLC Supplier No. HY14221A2MG
Siramesine hydrochloride, also known as Lu 28-179 hydrochloride, is a potent sigma-2 receptor agonist. It demonstrates high selectivity for sigma-2 receptors over sigma-1 receptors. This compound triggers cell death by destabilizing mitochondria and exhibits anti-cancer activity.
  • Potent sigma-2 receptor agonist
  • High selectivity for sigma-2 receptors
  • Triggers cell death via mitochondrial destabilization
  • Shows anti-cancer activity