Learn More
Cayman Chemical Siramesine (fumarate), Purity: ≥98%, CAS: 163630-79-3, 10 mg
SDP
Catalog No. 502986453
Encompass_Preferred
A selective agonist of σ2 receptor (IC50 = 0.19 nM); anxiolytic in animal studies and induces the death of cancer cells in vitro by destabilizing lysosomes
Catalog No. 50-298-6453 Supplier Cayman Chemical Supplier No. 2181710 MG
A selective agonist of σ2 receptor (IC50 = 0.19 nM); anxiolytic in animal studies and induces the death of cancer cells in vitro by destabilizing lysosomes