Learn More
MATTERHACKERS INC SIRAYA TRANSL BLUE LCD RSN 1L
SDP
Supplier:  MATTERHACKERS INC ML97AHR6
Encompass
Siraya Tech's Blu LCD Resin can be used to print strong, parts using an LCD 3D printer. When printing functional objects with just a bit of give and no brittle qualities, Blu Resin is the perfect fit. The optimal printing temp for Blu is over 25C, due to the material that provides its outstanding mechanical properties
Catalog No. NC2541893