Learn More
ILEX LIFE SCIENCES LLC Biocolor Sircol™ 2.0 Soluble Collagen Assay (96-well plate format)
SDP
Catalog No. NC2747063
Encompass_Preferred
The Biocolor Sircol™ 2.0 Soluble Collagen Assay is designed for quantitative measurement of soluble collagen in material from in-vivo and in-vitro sources. The kit employs colorimetric detection in a convenient microplate-based format.This latest addition to the highly cited family of "Sircol™" collagen kits combines an updated microplate format with an improved dye formulation to significantly enhance collagen sensitivity and specificity. This makes it possible to directly analyze collagen from complex samples such as conditioned media during 2D or 3D cell culture.
Catalog No. NC2747063 Supplier ILEX LIFE SCIENCES LLC Supplier No. SIRC2
The Biocolor Sircol™ 2.0 Soluble Collagen Assay is designed for quantitative measurement of soluble collagen in material from in-vivo and in-vitro sources. The kit employs colorimetric detection in a convenient microplate-based format.This latest addition to the highly cited family of "Sircol™" collagen kits combines an updated microplate format with an improved dye formulation to significantly enhance collagen sensitivity and specificity. This makes it possible to directly analyze collagen from complex samples such as conditioned media during 2D or 3D cell culture.