Learn More
BioVendor, LLC Sircol Acid-Salt Wash Reagent (3 x 20 ml), concentrated, Biocolor Item # SWASH
SDP
Catalog No. 501725968
Encompass
Sircol; Acid-Salt Wash Reagent (3 x 20 ml), concentrated. Contains acetic acid, sodium chloride and surfactants. Dilute the contents of the vial to 100 ml with deionised water before use. Sircol™ Collagen Assay component. Biocolor Item # SWASH
Catalog No. 50-172-5968 Supplier BioVendor, LLC Supplier No. SWASH
Sircol; Acid-Salt Wash Reagent (3 x 20 ml), concentrated. Contains acetic acid, sodium chloride and surfactants. Dilute the contents of the vial to 100 ml with deionised water before use. Sircol™ Collagen Assay component. Biocolor Item # SWASH