Learn More
ILEX LIFE SCIENCES LLC Biocolor Sircol Collagen Dye Reagent and Dye Release Reagent, Component Pack (2 x 110 ml Dye Reagent, 2 x 110 ml Dye Release Reagent)
SDP
Catalog No. 502120053
Encompass_Preferred
The Biocolor Sircol™ Soluble Collagen Assay is a quantitative assay for measurement of both acid-soluble and pepsin-soluble collagens. The colorimetric assay is a dye-binding method using Sircol™ Dye Reagent, which contains Sirius Red dye in picric acid. This kit is suitable for all soluble mammalian collagens, Types I to V. The assay can assess the rate of newly synthesized collagen. New collagen is produced during periods of rapid growth and development. New collagen is also generated during inflammation, wound healing and tumor development. The Sircol™ Assay can monitor collagen produced in situ, or during in-vitro cell culture. It can also measure in-vitro extracellular matrix (ECM) formation. Sensitivity: 1.0 µg. Assay Run Time: 1.5 hours. NOTE: This assay is not suitable for covalent cross-linked collagen.
Catalog No. 50-212-0053 Supplier ILEX LIFE SCIENCES LLC Supplier No. S1500
The Biocolor Sircol™ Soluble Collagen Assay is a quantitative assay for measurement of both acid-soluble and pepsin-soluble collagens. The colorimetric assay is a dye-binding method using Sircol™ Dye Reagent, which contains Sirius Red dye in picric acid. This kit is suitable for all soluble mammalian collagens, Types I to V. The assay can assess the rate of newly synthesized collagen. New collagen is produced during periods of rapid growth and development. New collagen is also generated during inflammation, wound healing and tumor development. The Sircol™ Assay can monitor collagen produced in situ, or during in-vitro cell culture. It can also measure in-vitro extracellular matrix (ECM) formation. Sensitivity: 1.0 µg. Assay Run Time: 1.5 hours. NOTE: This assay is not suitable for covalent cross-linked collagen.