Learn More
ILEX LIFE SCIENCES LLC SIRCOL ISO-CONC RGT 3 X 20ML
SDP
Catalog No. 502733619
Encompass_Preferred
Biocolor Sircol Isolation-Concentration Reagent (3 x 20 ml), component for use with Sircol Soluble Collagen Assay. The Biocolor Sircol Soluble Collagen Assay is a quantitative assay for measurement of both acid-soluble and pepsin-soluble collagens. The colorimetric assay is a dye-binding method using Sircol Dye Reagent, which contains Sirius Red dye in picric acid. This kit is suitable for all soluble mammalian collagens, Types I to V. The assay can assess the rate of newly synthesized collagen. New collagen is produced during periods of rapid growth and development. New collagen is also generated during inflammation, wound healing and tumor development. The Sircol Assay can monitor collagen produced in situ, or during in-vitro cell culture. It can also measure in-vitro extracellular matrix (ECM) formation.
Catalog No. 50-273-3619 Supplier ILEX LIFE SCIENCES LLC Supplier No. SICR
Biocolor Sircol Isolation-Concentration Reagent (3 x 20 ml), component for use with Sircol Soluble Collagen Assay. The Biocolor Sircol Soluble Collagen Assay is a quantitative assay for measurement of both acid-soluble and pepsin-soluble collagens. The colorimetric assay is a dye-binding method using Sircol Dye Reagent, which contains Sirius Red dye in picric acid. This kit is suitable for all soluble mammalian collagens, Types I to V. The assay can assess the rate of newly synthesized collagen. New collagen is produced during periods of rapid growth and development. New collagen is also generated during inflammation, wound healing and tumor development. The Sircol Assay can monitor collagen produced in situ, or during in-vitro cell culture. It can also measure in-vitro extracellular matrix (ECM) formation.