Learn More
Chondrex Inc Sirius Red Total Collagen Detection Kit
Catalog No. NC0219811
Encompass_Preferred
Sirius Red Collagen Detection Kit can be used for various collagen-containing samples such as tissue specimens, cell culture medium, and cultured cells. The total assay time is less than 30 minutes for up to 40 samples in duplicate.
Catalog No. NC0219811 Supplier Chondrex Inc Supplier No. 9062
Sirius Red Collagen Detection Kit can be used for various collagen-containing samples such as tissue specimens, cell culture medium, and cultured cells. The total assay time is less than 30 minutes for up to 40 samples in duplicate.