Learn More
Cell Signaling Technology Signalsilence™ p38 MAPK siRNA I, Human, 300 μL
SDP
Catalog No. 50194008
Encompass
SignalSilence™ p38 MAP Kinase siRNA I enables specific inhibition of p38 MAP kinase expression through RNA interference. By introducing double-stranded RNA molecules into the cell, this product allows researchers to selectively silence target gene expression. Every lot is rigorously tested and validated to ensure a reduction in protein expression by western analysis.
  • Supplied as an annealed RNA duplex at a concentration of 10 μM.
  • Oligonucleotide synthesis monitored by trityl analysis to ensure coupling efficiency.
  • Purified by affinity-solid phase extraction.
  • Verified by mass spectrometry to confirm composition and lot-to-lot consistency.
  • Recommended for use in human cell transfection at a final concentration of 100 nM.
  • Supplied in RNAse-free water for stability.
Catalog No. 50-194-008 Supplier Cell Signaling Technology Supplier No. 6564S
SignalSilence™ p38 MAP Kinase siRNA I enables specific inhibition of p38 MAP kinase expression through RNA interference. By introducing double-stranded RNA molecules into the cell, this product allows researchers to selectively silence target gene expression. Every lot is rigorously tested and validated to ensure a reduction in protein expression by western analysis.
  • Supplied as an annealed RNA duplex at a concentration of 10 μM.
  • Oligonucleotide synthesis monitored by trityl analysis to ensure coupling efficiency.
  • Purified by affinity-solid phase extraction.
  • Verified by mass spectrometry to confirm composition and lot-to-lot consistency.
  • Recommended for use in human cell transfection at a final concentration of 100 nM.
  • Supplied in RNAse-free water for stability.