Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology SignalSilence™ Stat3 siRNA I, Human, 3 nmol
SDP

Catalog No. 50194016
Encompass

SignalSilence™ Stat3 siRNA I allows for the specific inhibition of Stat3 expression through RNA interference. This method enables selective gene silencing via the delivery of double-stranded RNA molecules into cells. Each product undergoes rigorous in-house testing and is validated to reduce target protein expression by western analysis.

  • Specifically targets STAT3
  • Validated for transfection applications
  • Oligonucleotide synthesis monitored via trityl analysis
  • Purified by affinity-solid phase extraction
  • Annealed RNA duplex verified by mass spectrometry
  • Species reactivity: Human

Catalog No. 50-194-016 Supplier Cell Signaling Technology Supplier No. 6580S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SignalSilence™ Stat3 siRNA I allows for the specific inhibition of Stat3 expression through RNA interference. This method enables selective gene silencing via the delivery of double-stranded RNA molecules into cells. Each product undergoes rigorous in-house testing and is validated to reduce target protein expression by western analysis.

  • Specifically targets STAT3
  • Validated for transfection applications
  • Oligonucleotide synthesis monitored via trityl analysis
  • Purified by affinity-solid phase extraction
  • Annealed RNA duplex verified by mass spectrometry
  • Species reactivity: Human

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.