Learn More
Cell Signaling Technology SignalSilence™ Ezh2 siRNA I, 3 nmol, 10 μM
SDP
Catalog No. 50193980
Encompass
SignalSilence™ Ezh2 siRNA I enables the specific inhibition of Ezh2 expression through RNA interference, allowing for the selective silencing of the target gene via the delivery of double-stranded RNA. This product is suitable for transfection experiments and has been validated to reduce target protein expression by western analysis.
  • Supplied as a 10 μM solution in RNAse-free water.
  • Validated for use in human species.
  • Purified by affinity-solid phase extraction.
  • Subjected to mass spectrometry analysis to verify composition and ensure lot-to-lot consistency.
  • Recommended for use at a final concentration of 100 nM for 48 to 72 hours prior to cell lysis.
  • Each vial provides enough material for 100 transfections at a 300 μL total volume per well in a 24-well plate.
  • Aliquot and store at -20°C.
Catalog No. 50-193-980 Supplier Cell Signaling Technology Supplier No. 6509S
SignalSilence™ Ezh2 siRNA I enables the specific inhibition of Ezh2 expression through RNA interference, allowing for the selective silencing of the target gene via the delivery of double-stranded RNA. This product is suitable for transfection experiments and has been validated to reduce target protein expression by western analysis.
  • Supplied as a 10 μM solution in RNAse-free water.
  • Validated for use in human species.
  • Purified by affinity-solid phase extraction.
  • Subjected to mass spectrometry analysis to verify composition and ensure lot-to-lot consistency.
  • Recommended for use at a final concentration of 100 nM for 48 to 72 hours prior to cell lysis.
  • Each vial provides enough material for 100 transfections at a 300 μL total volume per well in a 24-well plate.
  • Aliquot and store at -20°C.