Learn More
Cell Signaling Technology SignalSilence™ hnRNP A1 siRNA I, 3 nmol
SDP
Catalog No. 50194460
Encompass
SignalSilence™ hnRNP A1 siRNA I enables the specific inhibition of hnRNP A1 expression via RNA interference, a method for selective gene silencing through the introduction of double-stranded RNA molecules into cells. This siRNA reagent is validated for its ability to reduce target protein expression, as confirmed by western analysis.
  • Inhibits human, mouse, rat, and monkey hnRNP A1 expression
  • Tested in-house for reliable performance
  • Provided in RNase-free water
  • Suitable for 100 transfections at a final concentration of 100 nM in a 24-well plate format
  • Rigorous quality control via mass spectrometry to ensure lot-to-lot consistency
  • Oligonucleotide synthesis verified by trityl analysis and purified by affinity-solid phase extraction
Catalog No. 50-194-460 Supplier Cell Signaling Technology Supplier No. 7668S
SignalSilence™ hnRNP A1 siRNA I enables the specific inhibition of hnRNP A1 expression via RNA interference, a method for selective gene silencing through the introduction of double-stranded RNA molecules into cells. This siRNA reagent is validated for its ability to reduce target protein expression, as confirmed by western analysis.
  • Inhibits human, mouse, rat, and monkey hnRNP A1 expression
  • Tested in-house for reliable performance
  • Provided in RNase-free water
  • Suitable for 100 transfections at a final concentration of 100 nM in a 24-well plate format
  • Rigorous quality control via mass spectrometry to ensure lot-to-lot consistency
  • Oligonucleotide synthesis verified by trityl analysis and purified by affinity-solid phase extraction