Learn More
Cell Signaling Technology SignalSilence™ SAPK/JNK siRNA I, 300 μL
SDP
Catalog No. 50193808
Encompass
SignalSilence™ SAPK/JNK siRNA I enables the specific inhibition of SAPK/JNK expression through RNA interference, a process that selectively silences gene expression via the delivery of double-stranded RNA molecules. This siRNA product is rigorously tested to ensure a reduction in target protein expression as validated by western analysis.
  • Inhibits JNK1 and JNK2 expression in human, mouse, rat, and monkey samples
  • Supplied in RNAse-free water
  • Verified lot-to-lot consistency via mass spectrometry
  • Validated for transfection applications
Catalog No. 50-193-808 Supplier Cell Signaling Technology Supplier No. 6232S
SignalSilence™ SAPK/JNK siRNA I enables the specific inhibition of SAPK/JNK expression through RNA interference, a process that selectively silences gene expression via the delivery of double-stranded RNA molecules. This siRNA product is rigorously tested to ensure a reduction in target protein expression as validated by western analysis.
  • Inhibits JNK1 and JNK2 expression in human, mouse, rat, and monkey samples
  • Supplied in RNAse-free water
  • Verified lot-to-lot consistency via mass spectrometry
  • Validated for transfection applications