Learn More
Cell Signaling Technology SignalSilence™ p44/42 MAPK siRNA, Human, 300 μL
SDP
Catalog No. 50194006
Encompass
SignalSilence™ p44/42 MAPK (Erk1/2) siRNA allows researchers to specifically inhibit p44/42 MAP Kinase expression via RNA interference, a method for selective silencing of gene expression through the delivery of double-stranded RNA molecules. This product is supplied as an aqueous solution and has been rigorously tested in-house to reduce protein expression as confirmed by western analysis.
  • Specifically inhibits p44/42 MAP Kinase expression
  • Validated by western analysis for protein knockdown efficiency
  • Supplied in RNAse-free water
  • Suitable for transfection
  • Each vial contains enough material for 100 transfections at a 100 nM concentration
Catalog No. 50-194-006 Supplier Cell Signaling Technology Supplier No. 6560S
SignalSilence™ p44/42 MAPK (Erk1/2) siRNA allows researchers to specifically inhibit p44/42 MAP Kinase expression via RNA interference, a method for selective silencing of gene expression through the delivery of double-stranded RNA molecules. This product is supplied as an aqueous solution and has been rigorously tested in-house to reduce protein expression as confirmed by western analysis.
  • Specifically inhibits p44/42 MAP Kinase expression
  • Validated by western analysis for protein knockdown efficiency
  • Supplied in RNAse-free water
  • Suitable for transfection
  • Each vial contains enough material for 100 transfections at a 100 nM concentration