Learn More
Cell Signaling Technology SignalSilence™ Akt siRNA I, Human, Mouse, Rat, 3 nmol
SDP
Catalog No. 50193791
Encompass
SignalSilence™ Akt siRNA I allows researchers to specifically inhibit Akt expression using RNA interference, a method where gene expression is selectively silenced via the delivery of double-stranded RNA molecules into the cell. Each lot is rigorously tested to ensure target protein expression reduction via Western analysis.
  • Inhibits human, mouse, and rat Akt1 and Akt2 expression
  • Does not affect Akt3 expression
  • Supplied in RNAse-free water
  • Validated by mass spectrometry for composition and lot-to-lot consistency
  • Designed for 100 transfections at a final concentration of 100 nM per transfection
Catalog No. 50-193-791 Supplier Cell Signaling Technology Supplier No. 6211S
SignalSilence™ Akt siRNA I allows researchers to specifically inhibit Akt expression using RNA interference, a method where gene expression is selectively silenced via the delivery of double-stranded RNA molecules into the cell. Each lot is rigorously tested to ensure target protein expression reduction via Western analysis.
  • Inhibits human, mouse, and rat Akt1 and Akt2 expression
  • Does not affect Akt3 expression
  • Supplied in RNAse-free water
  • Validated by mass spectrometry for composition and lot-to-lot consistency
  • Designed for 100 transfections at a final concentration of 100 nM per transfection