Learn More
Santa Cruz Biotechnology siRNA Dilution Buffer, 1.5 mL
Catalog No. NC9657127
Encompass
TRIS-EDTA based buffer prepared from RNase-free water suitable for storage and dilution of siRNA, pH 8.
Catalog No. NC9657127 Supplier Santa Cruz Biotechnology Supplier No. SC29527
TRIS-EDTA based buffer prepared from RNase-free water suitable for storage and dilution of siRNA, pH 8.