Learn More
Medchemexpress LLC SiRNA Negative Control | RNA, (UUCUCCGAACGUGUCACGUUU), complex with RNA, (ACGUGACACGUUCGGAGAAUU) (1:1) | 1 MG
SDP
Catalog No. 5000374120
Encompass_Preferred
SiRNA Negative Control is a siRNA of 21 nucleotides that can be used as a negative control. It has no homology to most known gene sequences and is suitable for use in human, mouse, and rat cells in vitro. This control serves as an experimental benchmark to verify reliability and act as a standardization reference, making it a common choice in many research articles.
  • SiRNA of 21 nucleotides
  • Can be used as a negative control
  • Has no homology to most known gene sequences
  • Can be used in human, mouse, and rat cells in vitro
  • Serves as an experimental control benchmark
  • Verifies experimental reliability
  • Acts as a standardization reference
  • Commonly used in most research articles
Catalog No. 50-003-74120 Supplier Medchemexpress LLC Supplier No. HY1501501MG
SiRNA Negative Control is a siRNA of 21 nucleotides that can be used as a negative control. It has no homology to most known gene sequences and is suitable for use in human, mouse, and rat cells in vitro. This control serves as an experimental benchmark to verify reliability and act as a standardization reference, making it a common choice in many research articles.
  • SiRNA of 21 nucleotides
  • Can be used as a negative control
  • Has no homology to most known gene sequences
  • Can be used in human, mouse, and rat cells in vitro
  • Serves as an experimental control benchmark
  • Verifies experimental reliability
  • Acts as a standardization reference
  • Commonly used in most research articles