Learn More
Waters Corp Waters Protein Precipitation Plate, 96-well, Polypropylene, Mass Spec Compatible
Catalog No. 50782047 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Protein Precipitation Plate enables high-throughput, in-well protein precipitation of biological samples. It prevents cloudy filtrates and clogged devices, optimizing performance by reducing processing steps. Ensures complete recovery of clean filtrate from small plasma volumes for drug metabolism, ADME, and toxicology labs. Yields precipitate-free filtrates for LC/MS analysis.
  • High-throughput, in-well protein precipitation.
  • Unique filter system & patented valve technology.
  • Prevents leakage, splashing, & cross-talk.
  • Filters via vacuum or centrifugation (2,000 × g, 1 min).
  • Maximizes sample recovery, reduces cross-talk, boosts reliability.
  • Removes hazy filtrates; maximizes Mass Spec uptime.
  • Manual or automated processing.
  • Clear filtrate for LC/MS autosampler or storage.
Catalog No. 50-782-047 Supplier Waters Corp Supplier No. 186003873
This Protein Precipitation Plate enables high-throughput, in-well protein precipitation of biological samples. It prevents cloudy filtrates and clogged devices, optimizing performance by reducing processing steps. Ensures complete recovery of clean filtrate from small plasma volumes for drug metabolism, ADME, and toxicology labs. Yields precipitate-free filtrates for LC/MS analysis.
  • High-throughput, in-well protein precipitation.
  • Unique filter system & patented valve technology.
  • Prevents leakage, splashing, & cross-talk.
  • Filters via vacuum or centrifugation (2,000 × g, 1 min).
  • Maximizes sample recovery, reduces cross-talk, boosts reliability.
  • Removes hazy filtrates; maximizes Mass Spec uptime.
  • Manual or automated processing.
  • Clear filtrate for LC/MS autosampler or storage.