Learn More
Medchemexpress LLC Sirolimus-13C,d3 | 00-00-0 | MFCD00867594 | 97.1% | 918.18 g/mol | C5013CH76D3NO13 | 1 MG
SDP
Catalog No. 5000246726
Encompass_Preferred
Rapamycin-13C,d3 is a stable isotope-labeled form of rapamycin supplied as a high-purity reference standard for use as an internal standard or tracer in quantitative bioanalytical methods such as LC-MS and GC-MS. It preserves the chemical identity of rapamycin while incorporating 13C and deuterium labels to aid accurate quantitation and metabolic studies.
  • Stable isotope-labeled (13C and d3) for accurate quantitation.
  • Suitable as an internal standard for LC-MS and GC-MS workflows.
  • High purity (97.05%) supports reliable analytical performance.
  • Available in small analytical pack sizes (500 μg, 1 mg, 5 mg, 10 mg).
  • Storage recommendations: -20°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month).
Catalog No. 50-002-46726 Supplier Medchemexpress LLC Supplier No. HY10219S11MG
Rapamycin-13C,d3 is a stable isotope-labeled form of rapamycin supplied as a high-purity reference standard for use as an internal standard or tracer in quantitative bioanalytical methods such as LC-MS and GC-MS. It preserves the chemical identity of rapamycin while incorporating 13C and deuterium labels to aid accurate quantitation and metabolic studies.
  • Stable isotope-labeled (13C and d3) for accurate quantitation.
  • Suitable as an internal standard for LC-MS and GC-MS workflows.
  • High purity (97.05%) supports reliable analytical performance.
  • Available in small analytical pack sizes (500 μg, 1 mg, 5 mg, 10 mg).
  • Storage recommendations: -20°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month).