Learn More
Medchemexpress LLC Sirolimus | 53123-88-9 | MFCD00867594 | 914.2 g/mol | C51H79NO13 | 10 MG
SDP
Catalog No. 5000245829
Encompass_Preferred
Rapamycin (sirolimus) GMP-grade is a macrolide mTOR inhibitor supplied for use as a GMP reagent in regulated research and cell therapy workflows. It binds the immunophilin FKBP12 to specifically inhibit mTORC1, and is reported to act as an immunosuppressant and autophagy activator. Supplied in a small 10 mg pack appropriate for controlled-process applications.
  • GMP-grade material intended for regulated workflows.
  • Potent mTOR inhibitor with reported IC50 ~0.1 nM in HEK293 cells.
  • Acts as an immunosuppressant and autophagy activator.
  • Provided in small pack sizes suitable for cell therapy reagent use.
Catalog No. 50-002-45829 Supplier Medchemexpress LLC Supplier No. HY10219G10MG
Rapamycin (sirolimus) GMP-grade is a macrolide mTOR inhibitor supplied for use as a GMP reagent in regulated research and cell therapy workflows. It binds the immunophilin FKBP12 to specifically inhibit mTORC1, and is reported to act as an immunosuppressant and autophagy activator. Supplied in a small 10 mg pack appropriate for controlled-process applications.
  • GMP-grade material intended for regulated workflows.
  • Potent mTOR inhibitor with reported IC50 ~0.1 nM in HEK293 cells.
  • Acts as an immunosuppressant and autophagy activator.
  • Provided in small pack sizes suitable for cell therapy reagent use.