Learn More
Medchemexpress LLC Sirolimus | 53123-88-9 | MFCD00867594 | 99.0% | 914.17 g/mol | C51H79NO13 | 50 MG
SDP
Catalog No. 5000247064
Encompass_Preferred
Analytical standard of rapamycin (sirolimus) supplied as a solid reference material for research and analytical use. It provides defined identity and high purity for assay calibration, method development, and quality control.
  • Analytical standard of sirolimus (rapamycin).
  • High purity (98.98%) suitable for analytical applications.
  • Molecular formula C51H79NO13; molecular weight 914.17 g/mol.
  • Provided as a solid powder for laboratory use.
  • Available in a 50 MG pack size for analytical workflows.
Catalog No. 50-002-47064 Supplier Medchemexpress LLC Supplier No. HY10219R50MG
Analytical standard of rapamycin (sirolimus) supplied as a solid reference material for research and analytical use. It provides defined identity and high purity for assay calibration, method development, and quality control.
  • Analytical standard of sirolimus (rapamycin).
  • High purity (98.98%) suitable for analytical applications.
  • Molecular formula C51H79NO13; molecular weight 914.17 g/mol.
  • Provided as a solid powder for laboratory use.
  • Available in a 50 MG pack size for analytical workflows.