Learn More
MOLECULAR DEPOT LLC Sirolimus BSA - 0.5 mg
Catalog No. 5000013585
Encompass_Preferred
Sirolimus BSA Conjugate Solution is a concentrated solution of Bovine Serum Albumin (BSA) conjugated to Sirolimus A carbon spacer arm links BSA to the Sirolimus moiety Each BSA molecule contains several Sirolimus units Dilutions of the provided conjugate can be made with Phosphate Buffer Saline For in vitro research use only
Catalog No. 50-000-13585 Supplier MOLECULAR DEPOT LLC Supplier No. P2010025
Sirolimus BSA Conjugate Solution is a concentrated solution of Bovine Serum Albumin (BSA) conjugated to Sirolimus A carbon spacer arm links BSA to the Sirolimus moiety Each BSA molecule contains several Sirolimus units Dilutions of the provided conjugate can be made with Phosphate Buffer Saline For in vitro research use only